Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps9d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079393.1 | NP_001072861.1 | 10 - 11 | 1447 | 88 | |
GenBank mRNA | BC123082.1 | - | 10 - 11 | 1447 | 88 | |
CR848484.2 | - | 10 - 11 | 1484 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |