Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: znrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079396.1 | NP_001072864.1 | 7 - 8 | 1259 | 78 | |
XM_012966110.2 | XP_012821564.1 | 6 - 7 | 4709 | 78 | ||
GenBank mRNA | BC123917.1 | - | 7 - 8 | 1259 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |