Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: neto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079444.1 | NP_001072912.1 | 5 - 6 | 852 | 81 | |
GenBank mRNA | BC121922.1 | - | 5 - 6 | 852 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |