Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079453.1 | NP_001072921.1 | 8 - 9 | 808 | 98 | |
XM_012959324.2 | XP_012814778.1 | 6 - 7 | 1823 | 98 | ||
XM_018092190.1 | XP_017947679.1 | 8 - 9 | 823 | 98 | ||
GenBank mRNA | BC121460.1 | - | 8 - 9 | 808 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |