Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc2a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079458.1 | NP_001072926.1 | 4 - 5 | 1577 | 71 | |
XM_012952588.2 | XP_012808042.1 | 4 - 5 | 1885 | 71 | ||
XM_012952589.2 | XP_012808043.1 | 4 - 5 | 1905 | 71 | ||
XM_012952590.2 | XP_012808044.1 | 5 - 6 | 1769 | 71 | ||
XM_012952592.2 | XP_012808046.1 | 4 - 5 | 1615 | 71 | ||
XM_012952593.2 | XP_012808047.1 | 4 - 5 | 1613 | 71 | ||
XM_012952595.2 | XP_012808049.1 | 4 - 5 | 1608 | 71 | ||
XM_012952596.2 | XP_012808050.1 | 4 - 5 | 1930 | 71 | ||
GenBank mRNA | BC122089.1 | - | 4 - 5 | 1577 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |