Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc28b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016093.3 | NP_001016093.1 | 4 - 5 | 623 | 83 | |
XM_012956925.2 | XP_012812379.1 | 4 - 5 | 572 | 83 | ||
XM_012956926.2 | XP_012812380.1 | 4 - 5 | 644 | 83 | ||
XM_018091237.1 | XP_017946726.1 | 3 - 4 | 574 | 83 | ||
GenBank mRNA | BC121527.1 | - | 4 - 5 | 623 | 83 | |
CR762178.2 | - | 4 - 5 | 740 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |