Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: derl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016097.3 | NP_001016097.1 | 5 - 6 | 535 | 111 | |
GenBank mRNA | BC135880.1 | - | 5 - 6 | 535 | 111 | |
CR762155.2 | - | 5 - 6 | 554 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |