Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016125.3 | NP_001016125.1 | 2 - 3 | 138 | 82 | |
GenBank mRNA | BC128617.1 | - | 2 - 3 | 138 | 82 | |
CR762066.4 | - | 2 - 3 | 151 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |