Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017068.3 | NP_001017068.1 | 1 - 2 | 111 | 80 | |
XM_012957025.2 | XP_012812479.1 | 1 - 2 | 224 | 80 | ||
GenBank mRNA | BC135652.1 | - | 1 - 2 | 111 | 80 | |
CR761572.2 | - | 1 - 2 | 101 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |