Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ebag9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017088.3 | NP_001017088.1 | 3 - 4 | 317 | 100 | |
XM_012964771.2 | XP_012820225.1 | 4 - 5 | 411 | 100 | ||
XM_012964772.2 | XP_012820226.1 | 3 - 4 | 331 | 100 | ||
XM_012964773.2 | XP_012820227.1 | 4 - 5 | 404 | 100 | ||
GenBank mRNA | BC135192.1 | - | 3 - 4 | 317 | 100 | |
CR761412.2 | - | 2 - 3 | 174 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |