Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dusp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045578.2 | NP_001039043.2 | 2 - 3 | 1140 | 83 | |
GenBank mRNA | BC118778.1 | - | 2 - 3 | 970 | 83 | |
BC135306.1 | - | 2 - 3 | 1140 | 83 | ||
CR848381.2 | - | 2 - 3 | 979 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |