Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: opcml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079019.1 | NP_001072487.1 | 7 - 8 | 1005 | 94 | |
GenBank mRNA | BC121914.1 | - | 7 - 8 | 1005 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |