Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rims3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079023.1 | NP_001072491.1 | 5 - 6 | 617 | 70 | |
GenBank mRNA | BC121920.1 | - | 5 - 6 | 617 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |