Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079044.1 | NP_001072512.1 | 12 - 13 | 1400 | 88 | |
GenBank mRNA | BC121953.1 | - | 12 - 13 | 1400 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |