Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079044.1 | NP_001072512.1 | 22 - 23 | 3279 | 96 | |
GenBank mRNA | BC121953.1 | - | 22 - 23 | 3279 | 96 |
Gene Symbol: LOC108647660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018094188.1 | XP_017949677.1 | 2 - 3 | 622 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |