Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079094.1 | NP_001072562.1 | 3 - 4 | 251 | 88 | |
GenBank mRNA | BC122084.1 | - | 3 - 4 | 251 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |