Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079119.1 | NP_001072587.1 | 1 - 2 | 865 | 71 | |
XM_012970558.2 | XP_012826012.1 | 1 - 2 | 898 | 71 | ||
GenBank mRNA | BC122952.1 | - | 1 - 2 | 865 | 71 | |
BC136086.1 | - | 1 - 2 | 737 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |