Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079267.1 | NP_001072735.1 | 7 - 8 | 612 | 98 | |
XM_012968693.2 | XP_012824147.1 | 8 - 9 | 866 | 98 | ||
XM_012968694.2 | XP_012824148.1 | 7 - 8 | 830 | 98 | ||
GenBank mRNA | BC125715.1 | - | 7 - 8 | 612 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |