Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: car1_predicted | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079317.1 | NP_001072785.1 | 6 - 7 | 678 | 101 | |
XM_012964940.2 | XP_012820394.1 | 6 - 7 | 738 | 101 | ||
XM_012964941.2 | XP_012820395.1 | 6 - 7 | 813 | 101 | ||
XM_012964943.2 | XP_012820397.1 | 6 - 7 | 1150 | 101 | ||
XM_012964944.2 | XP_012820398.1 | 6 - 7 | 732 | 101 | ||
XM_018094705.1 | XP_017950194.1 | 6 - 7 | 1697 | 101 | ||
GenBank mRNA | BC118911.1 | - | 6 - 7 | 678 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |