Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prdm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079430.1 | NP_001072898.1 | 3 | 669 | 81 | |
GenBank mRNA | BC118858.1 | - | 3 | 669 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Both primers and probe map within a single exon |