Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079433.1 | NP_001072901.1 | 3 - 4 | 295 | 61 | |
GenBank mRNA | BC121430.1 | - | 3 - 4 | 295 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |