Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npas4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079438.2 | NP_001072906.2 | 6 - 7 | 1043 | 74 | |
XM_012961436.2 | XP_012816890.1 | 6 - 7 | 1863 | 74 | ||
GenBank mRNA | BC121975.1 | - | 5 - 6 | 1498 | 74 | |
BC136031.1 | - | 6 - 7 | 1043 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |