Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klhl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079442.1 | NP_001072910.1 | 3 - 4 | 545 | 87 | |
XM_012966300.2 | XP_012821754.1 | 3 - 4 | 592 | 87 | ||
XM_012966303.2 | XP_012821757.1 | 3 - 4 | 577 | 87 | ||
XM_012966306.2 | XP_012821760.1 | 3 - 4 | 532 | 87 | ||
XM_012966310.2 | XP_012821764.1 | 3 - 4 | 543 | 87 | ||
XM_012966315.2 | XP_012821769.1 | 2 - 3 | 238 | 87 | ||
GenBank mRNA | BC124036.1 | - | 3 - 4 | 545 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |