Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083354.2 | NP_001076823.1 | 3 - 4 | 687 | 66 | |
XM_012959159.2 | XP_012814613.1 | 3 - 4 | 1216 | 66 | ||
GenBank mRNA | BC135750.1 | - | 3 - 4 | 687 | 66 | |
CR761177.2 | - | 3 - 4 | 634 | 66 | ||
CT485733.1 | - | 3 - 4 | 639 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |