Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bckdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097214.1 | NP_001090683.1 | 3 - 4 | 606 | 72 | |
XM_012967799.2 | XP_012823253.1 | 5 - 6 | 828 | 72 | ||
GenBank mRNA | BC127357.1 | - | 3 - 4 | 606 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |