Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bckdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097214.1 | NP_001090683.1 | 4 - 5 | 815 | 82 | |
XM_012967799.2 | XP_012823253.1 | 6 - 7 | 1037 | 82 | ||
GenBank mRNA | BC127357.1 | - | 4 - 5 | 815 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |