Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmeff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097223.2 | NP_001090692.1 | 6 - 7 | 675 | 86 | |
XM_012969926.2 | XP_012825380.1 | 7 - 8 | 1044 | 86 | ||
GenBank mRNA | BC127375.1 | - | 6 - 7 | 675 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |