Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmeff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097223.2 | NP_001090692.1 | 9 - 10 | 1060 | 83 | |
GenBank mRNA | BC127375.1 | - | 9 - 10 | 1060 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |