Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dipk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097225.1 | NP_001090694.1 | 411 | 71 | ||
GenBank mRNA | BC127379.1 | - | 411 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis |