Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dipk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097225.1 | NP_001090694.1 | 2 - 3 | 821 | 103 | |
GenBank mRNA | BC127379.1 | - | 2 - 3 | 821 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |