Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chsy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097255.1 | NP_001090724.1 | 1 - 2 | 863 | 101 | |
GenBank mRNA | BC127293.1 | - | 1 - 2 | 863 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |