Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st8sia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097280.1 | NP_001090749.1 | 6 - 7 | 595 | 78 | |
GenBank mRNA | AM422137.1 | - | 6 - 7 | 595 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Xenopus tropicalis Probe spans exons |