Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097281.1 | NP_001090750.1 | 453 | 96 | ||
GenBank mRNA | AM262832.1 | - | 453 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis |