Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097318.1 | NP_001090787.1 | 6 - 7 | 675 | 94 | |
XM_012960380.2 | XP_012815834.1 | 6 - 7 | 741 | 94 | ||
XM_012960381.2 | XP_012815835.1 | 5 - 6 | 692 | 94 | ||
GenBank mRNA | BC135170.1 | - | 6 - 7 | 675 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |