Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: caps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097320.1 | NP_001090789.1 | 4 - 5 | 598 | 98 | |
XM_012957609.2 | XP_012813063.1 | 5 - 6 | 657 | 98 | ||
GenBank mRNA | BC135179.1 | - | 4 - 5 | 598 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |