Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097344.1 | NP_001090813.1 | 4 - 5 | 1158 | 69 | |
GenBank mRNA | BC135260.1 | - | 4 - 5 | 1158 | 69 | |
HQ018040.1 | - | 4 - 5 | 1092 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |