Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097347.1 | NP_001090816.1 | 3 - 4 | 812 | 71 | |
GenBank mRNA | BC135267.1 | - | 3 - 4 | 812 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |