Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rfx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097380.1 | NP_001090849.1 | 1 - 2 | 588 | 69 | |
GenBank mRNA | BC133720.1 | - | 1 - 2 | 588 | 69 | |
CR761154.2 | - | 1 - 2 | 353 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |