Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dipk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097382.1 | NP_001090851.1 | 4 - 5 | 391 | 63 | |
XM_012960387.2 | XP_012815841.1 | 5 - 6 | 727 | 63 | ||
XM_012960388.2 | XP_012815842.1 | 4 - 5 | 668 | 63 | ||
GenBank mRNA | BC135136.1 | - | 4 - 5 | 391 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |