Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097384.1 | NP_001090853.1 | 4 - 5 | 790 | 82 | |
GenBank mRNA | BC135141.1 | - | 4 - 5 | 790 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |