Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slx4ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097385.1 | NP_001090854.1 | 4 - 5 | 335 | 103 | |
XM_012962569.2 | XP_012818023.1 | 4 - 5 | 364 | 103 | ||
XM_012962570.2 | XP_012818024.1 | 3 - 4 | 275 | 103 | ||
XM_018093741.1 | XP_017949230.1 | 5 - 6 | 469 | 103 | ||
GenBank mRNA | BC135156.1 | - | 4 - 5 | 335 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |