Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: esrrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100210.1 | NP_001093680.1 | 8 - 9 | 1475 | 100 | |
XM_012962574.2 | XP_012818028.1 | 7 - 8 | 1392 | 100 | ||
XM_012962575.2 | XP_012818029.1 | 6 - 7 | 1290 | 100 | ||
XM_012962576.2 | XP_012818030.1 | 7 - 8 | 1395 | 100 | ||
XM_012962577.2 | XP_012818031.1 | 6 - 7 | 1313 | 100 | ||
XM_012962578.1 | XP_012818032.1 | 8 - 9 | 1475 | 100 | ||
XM_012962579.2 | XP_012818033.1 | 7 - 8 | 1443 | 100 | ||
XM_012962580.2 | XP_012818034.1 | 6 - 7 | 1317 | 100 | ||
XM_018093742.1 | XP_017949231.1 | 7 - 8 | 1368 | 100 | ||
XM_018093743.1 | XP_017949232.1 | 7 - 8 | 1226 | 100 | ||
GenBank mRNA | BC136151.1 | - | 8 - 9 | 1475 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |