Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100228.1 | NP_001093698.1 | 3 - 4 | 1295 | 73 | |
GenBank mRNA | BC135731.1 | - | 3 - 4 | 1295 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |