Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sox14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100233.1 | NP_001093703.1 | 1 | 646 | 69 | |
GenBank mRNA | AM922298.1 | - | 1 | 597 | 69 | |
BC135637.1 | - | 1 | 646 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |