Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: igfbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100237.1 | NP_001093707.1 | 1 - 2 | 687 | 100 | |
GenBank mRNA | BC135928.1 | - | 1 - 2 | 687 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |