Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fscn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100254.1 | NP_001093724.1 | 3 - 4 | 1168 | 100 | |
GenBank mRNA | BC135752.1 | - | 3 - 4 | 1168 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |