Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dmrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100256.1 | NP_001093726.1 | 3 - 4 | 785 | 90 | |
GenBank mRNA | BC135447.1 | - | 3 - 4 | 785 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |