Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100258.1 | NP_001093728.1 | 3 - 4 | 1679 | 67 | |
GenBank mRNA | BC135445.1 | - | 3 - 4 | 1679 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |