Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfrsf11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012965431.2 | XP_012820885.1 | 8 - 9 | 1337 | 72 | |
GenBank mRNA | BC136133.1 | - | 7 - 8 | 1620 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |