Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmeff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100268.3 | NP_001093738.2 | 4 - 5 | 952 | 68 | |
XM_012964170.2 | XP_012819624.1 | 4 - 5 | 1002 | 68 | ||
GenBank mRNA | BC135422.2 | - | 4 - 5 | 786 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |